Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ly-49E/F Monoclonal Antibody (CM4), PerCP-eFluor™ 710, eBioscience™
GREENER_CHOICE

Catalog No. 5016086
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-160-86 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-160-86 Supplier Invitrogen™ Supplier No. 46584882
Add to Cart
Edge
Add to Cart

Rat Monoclonal Antibody

Description: This CM4 monoclonal antibody reacts with a subset of mouse Ly49 receptors, specifically Ly49E and Ly49F. The Ly49 family of receptors are clustered in the Natural Killer Cell complex on chromosome 6. Expression is confined to NK cell, lymphocytes that play a key role in protection from infection and malignant diseases as well as graft rejection and autoimmune diseases. Ly49E is unique from other family members in that it contains an ITIM (immunoreceptor tyrosine-based inhibitory motif) domain and is expressed on all mouse strains. Developmentally Ly49E is one of the first markers detected on NK cells but expression decreases after birth to about 1% of NK cells in the adult spleen. Upon stimulation, expression can again be induced on Ly49E negative cells. The CM4 antibody is a useful reagent for the characterization of Ly49E expression when used in combination with other Ly49 antibodies such as 14B11 (Ly49C/I/F/H), 3D10 (Ly49C), YLI-90 (Ly49I), and A1 (Ly49A). Applications Reported: This CM4 antibody has been reported for use in flow cytometric analysis. Applications Tested: This CM4 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be caref...

The Ly49 family of receptors are clustered in the Natural Killer Cell complex on chromosome 6. Expression is confined to NK cell, lymphocytes that play a key role in protection from infection and malignant diseases as well as graft rejection and autoimmune diseases. Ly49E is unique from other family members in that it contains an ITIM (immunoreceptor tyrosine-based inhibitory motif) domain and is expressed on all mouse strains. Developmentally Ly49E is one of the first markers detected on NK cells but expression decreases after birth to about 1% of NK cells in the adult spleen. Upon stimulation, expression can again be induced on Ly49E negative cells. The CM4 antibody is a useful reagent for the characterization of Ly49E expression when used in combination with other Ly49 antibodies such as 14B11 (Ly49C/I/F/H), 3D10 (Ly49C), YLI-90 (Ly49I), and A1 (Ly49A).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ly-49E/F
Applications Flow Cytometry
Classification Monoclonal
Clone CM4
Concentration 0.2 mg/mL
Conjugate PerCP-eFluor 710
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Klra5
Gene Accession No. Q60652, Q60653
Gene Alias class I MHC receptor KLRA5; killer cell lectin-like receptor 5; Killer cell lectin-like receptor 6; killer cell lectin-like receptor, subfamily A, member 5; killer cell lectin-like receptor, subfamily A, member 6; Klra5; Klra6; Ly49e; Ly-49e; Ly49-e; Ly49E receptor; Ly49f; ly-49f; Ly49-f; Lymphocyte antigen 49e; lymphocyte antigen 49f; T-cell surface glycoprotein Ly-49E; T-cell surface glycoprotein Ly-49F
Gene Symbols Klra5, Klra6
Host Species Rat
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16636, 16637
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
I am unable to analyze my cells stained with eFluor Organic Dyes today. What options do I have?

Our options will depend on the samples you are analyzing.

If cell viability is not critical, you can store your stained samples at 4 degrees C or on ice overnight in the dark and analyze the following day.

For samples stained with eFluor organic fluorochromes, we recommend that cells be suspended in 100 uL of Flow Cytometry Staining Buffer (Cat. No. 00-4222) and 100 uL of eBioscience IC Fixation Buffer (Cat. No. 00-8222); samples can be incubated for up to 3 days at 4 degrees C in the dark. Alternatively, the 1-step Fix/Lyse Solution (Cat. No. 00-5333) can be used. This is a great option when working with whole blood but also works for other cell types.

Can the eFluor Organic fluorochromes be used for intracellular staining?

Yes, the eFluor Organic fluorochromes can be used for intracellular staining. The eFluor organic fluorochromes maintain bright signal and require minimal changes in compensation when fixed with eBioscience IC Fixation Buffer (Cat. No. 00-8222-49) and Permeabilization Buffer (Cat. No. 00-8333-56) or 1-step Fix/Lyse Solution (Cat. No. 00-5333-54, 00-5333-57) (as compared to live cells).

What buffers are compatible with the eFluor Organic fluorochromes and eVolve QDots?

The eFluor Organic fluorochromes and eVolve QDots can be used with flow staining buffers containing PBS and protein.

What is the difference between eFluor Organic Dyes and eVolve Qdots?

The eFluor Organic Dyes (eFluor 450, APC-eFluor 780, PerCP-eFluor 710, eFluor 710) are conventional fluorochromes. In contrast, the eVolve line of products are Quantum dots.

Are the eFluor Organic Dyes photo-labile?

As with other fluorochromes, we recommend minimal exposure to light to maintain optimal signal.

WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.