Learn More
Invitrogen™ Ly-49H Monoclonal Antibody (3D10), Biotin, eBioscience™

Description
The monoclonal antibody 3D10 recognizes Ly-49H, a member of the C-type lectin Ly-49 multigene family of receptors found on natural killer cells. Ly-49H, like Ly-49D, lacks the characteristic ITIM (immunoreceptor tyrosine-based inhibitory motif) and contains an arginine in the transmembrane domain implying these molecules act as activation receptors. Ly-49H is expressed in a subset of NK cells but not NKT (NK1.1CD3+) cells. Natural killer cells can express several Ly-49 proteins. It has been shown that there is preferential staining of Ly-49H with Ly-49D(Product # 12-5783-81) . Expression has been confirmed on C57BL/6 and NWNA but not Balb/c or DBA/2 mice. Other mice strains have not been tested. No crossreactivity to Ly-49A, C, D or G2 has been observed. Addition of monoclonal antibody 3D10 to cell cultures induces lysis of target cells. Applications Reported:This 3D10 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 3D10 antibody has been tested by flow cytometric analysis of C57Bl/6 spleen cells. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser...
Specifications
Specifications
| Antigen | Ly-49H |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 3D10 |
| Concentration | 0.5 mg/mL |
| Conjugate | Biotin |
| Formulation | PBS with 0.09% sodium azide; pH 7.2 |
| Gene | Klra8 |
| Gene Accession No. | Q60682 |
| Gene Alias | Cmv1; Cmv-1; cytomegalovirus resistance 1; killer cell lectin-like receptor 8; killer cell lectin-like receptor, subfamily A, member 8; Klra8; Ly49H; Ly-49h; Ly49-h; Ly49u>129>; Lymphocyte antigen 49h; T-cell surface glycoprotein Ly-49H |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.