Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ly-49H Monoclonal Antibody (3D10), PE-Cyanine7, eBioscience™
GREENER_CHOICE

Catalog No. 25588682
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-588-682 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 25-588-682 Supplier Invitrogen™ Supplier No. 25588682
  • $291.00
    $218.65 / Each of 1

    Save $72.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

The monoclonal antibody 3D10 recognizes Ly-49H, a member of the C-type lectin Ly-49 multigene family of receptors found on natural killer cells. Ly-49H, like Ly-49D, lacks the characteristic ITIM (immunoreceptor tyrosine-based inhibitory motif) and contains an arginine in the transmembrane domain implying these molecules act as activation receptors. Ly-49H is expressed in a subset of NK cells but not NKT (NK1.1CD3+) cells. Natural killer cells can express several Ly-49 proteins. It has been shown that there is preferential staining of Ly-49H with Ly-49D (Product # 12-5783). Expression has been confirmed on C57BL/6 and NWNA but not Balb/c or DBA/2 mice. Other mice strains have not been tested. No crossreactivity to Ly-49A, C, D or G2 has been observed. Addition of monoclonal antibody 3D10 to cell cultures induces lysis of target cells. Applications Reported: This 3D10 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 3D10 antibody has been tested by fow cytometric analyis of mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Light sensitivity: This tandem dye is sensitive to photo-induce...

Ly49H is a member of the C-type lectin Ly-49 multigene family of receptors found on natural killer cells. Ly49H, like Ly49D, lacks the characteristic ITAM (immunoreceptor tyrosine-based inhibitory motif) and contains an arginine in the transmembrane domain implying these molecules act as activation receptors. Ly49H is expressed in a subset of NK cells but not NKT (NK1.1/CD3+) cells. Natural killer cells can express several Ly49 proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ly-49H
Applications Flow Cytometry
Classification Monoclonal
Clone 3D10
Concentration 0.2 mg/mL
Conjugate PE-Cyanine7
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Klra8
Gene Accession No. Q60682
Gene Alias Cmv1; Cmv-1; cytomegalovirus resistance 1; killer cell lectin-like receptor 8; killer cell lectin-like receptor, subfamily A, member 8; Klra8; Ly49H; Ly-49h; Ly49-h; Ly49u>129>; Lymphocyte antigen 49h; T-cell surface glycoprotein Ly-49H
Gene Symbols Klra8
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16639
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.