Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ly-6G/Ly-6C Monoclonal Antibody (RB6-8C5), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 46593182
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-593-182 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 46-593-182 Supplier Invitrogen™ Supplier No. 46593182
  • $308.00
    $231.65 / Each of 1

    Save $76.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rat Monoclonal Antibody

Granulocytes are a category of white blood cells characterised by the presence of granules in their cytoplasm. They are also called polymorphonuclear leukocytes (PMN or PML) because of the varying shapes of the nucleus, which is usually lobed into three segments. In common parlance, the term polymorphonuclear leukocyte often refers specifically to neutrophil granulocytes, the most abundant of the granulocytes. Granulocytes or PMN are released from the bone marrow by the regulatory complement proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ly-6G/Ly-6C
Applications Flow Cytometry
Classification Monoclonal
Clone RB6-8C5
Concentration 0.2 mg/mL
Conjugate PerCP-eFluor 710
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Ly6c1
Gene Accession No. P0CW02, P0CW03, P35461
Gene Alias AA682074; AA959465; Gr1; Gr-1; Granulocyte Marker; Ly6c; Ly-6C; Ly-6C.2; Ly6c1; Ly-6C1; Ly6c2; Ly-6C2; Ly6g; Ly-6G; ly-6G.1; lymphocyte antigen 6 complex, locus C; lymphocyte antigen 6 complex, locus C1; lymphocyte antigen 6 complex, locus C2; lymphocyte antigen 6 complex, locus G; lymphocyte antigen 6C precursor (Ly-6C); lymphocyte antigen 6C1; Lymphocyte antigen 6C2; lymphocyte antigen 6G; Lymphocyte antigen Ly-6C; lymphocyte differentiation antigen; PML; PMN
Gene Symbols Ly6c1, Ly6c2, Ly6g
Host Species Rat
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100041546, 17067, 546644
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Product Line eBioscience
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.