Learn More
Cayman Chemical LY2157299, 700874-72-2, 10 mg
SDP
Supplier:  Cayman Chemical 1531210
Encompass_Preferred
A small molecule inhibitor of the TGF-β receptor type 1 kinase (IC50 = 56 nM) that has been used to study the role of TGF-β signaling in triple negative breast cancer cells and hepatocellular carcinoma cells
Catalog No. NC0843494
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.