Learn More
Selleck Chemical LLC LY2409881 5mg 946518-60-1
SDP
Supplier:  Selleck Chemical LLC S76975MG
Encompass_Preferred
LY2409881 is a potent and selective IKK2 inhibitor with IC50 of 30 nM, >10-fold selectivity over IKK1 and other common kinases. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5143
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.