Learn More
TARGETMOL CHEMICALS INC LY255283 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T2294610MG
Encompass_Preferred
Also available in 1 mL, 2 mg, 5 mg, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes.LY255283 is a specific antagonit of leukotriene B4 (LTB4) receptor. It inhibits the production of LTB(4) in human peripheral blood polymorphonuclear leukocytes (PMNL) and in monocytes activated by calcium ionophore A23187. Purity 99.45%
Catalog No. 50-287-7564
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.