Learn More
Apexbio Technology LLC LY2801653 1206799-15-6 100mg
SDP
Supplier:  Apexbio Technology LLC A3573100
Encompass_Preferred
LY2801653 (CAS 1206799-15-6) is an orally bioavailable small molecule inhibitor targeting the c-MET receptor tyrosine kinase with an IC50 of 2 nM c-MET also known as hepatocyte growth factor receptor (HGFR) mediates key cellular processes including proliferation survival and motility its dysregulation has been associated with tumor progression and metastasis LY2801653 acts as an ATP-competitive inhibitor of c-MET phosphorylation In vitro it fully inhibits HGF-induced cell scattering in DU-145 cells at concentrations from 0 01 to 10 M and shows greater antiproliferative effects in MET-expressing cell lines In vivo LY2801653 reduces apoptosis and angiogenesis in xenograft models supporting its utility in cancer research
Catalog No. 50-000-93739
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.