Learn More
Apexbio Technology LLC LY2835219, 25mg. Cas: 1231930-82-7 MFCD: MFCD25562906. CDK4/6 inhibitor,potent and selective
SDP
Supplier:  Apexbio Technology LLC A179425
Encompass_Preferred
LY2835219 is a selective and orally available dual cyclin-dependent kinases 4/6 (CDK4/6) inhibitor that potently inhibits the activities of CDK4 and CDK6 with the half maximal inhibition concentration IC50 values of 2 nM and 10 nM respectively [1]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4709
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.