Learn More
Selleck Chemical LLC LY2874455 5mg 1254473-64-7
SDP
Supplier:  Selleck Chemical LLC S70575MG
Encompass_Preferred
LY2874455 is a pan-FGFR inhibitor with IC50 of 2.8 nM, 2.6 nM, 6.4 nM, and 6 nM for FGFR1, FGFR2, FGFR3, and FGFR4, respectively, and also inhibits VEGFR2 activity with IC50 of 7 nM. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0880138
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.