Learn More
Apexbio Technology LLC LY2886721, 5mg. Cas: 1262036-50-9 MFCD: MFCD22124078. BACE inhibitor
SDP
Supplier:  Apexbio Technology LLC A84655
Encompass_Preferred
LY2886721 is an oral and furothiazine-based inhibitor of site-amyloid protein cleaving enzyme (BACE) with IC50 of 20.3nM. This product has the potency for the treatment of Alzheimer's disease. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5148
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.