Learn More
Cayman Chemical LY294002, 154447-36-6, 5 mg
SDP
Supplier:  Cayman Chemical 709205
Encompass_Preferred
Molecular Formula C19H17NO3, Purity ≥98%, Formula Weight 307.3; A selective PI3K inhibitor with an IC50 value of 1.4 M.
Catalog No. NC9642062
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.