Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LY6G6D Recombinant Human Monoclonal Antibody (8I8)
GREENER_CHOICE

Catalog No. FEMA550088 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550088 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550088 Supplier Invitrogen™ Supplier No. MA550088
  • $505.00
    $446.65 / Each of 1

    Save $58.35

    12% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Human Recombinant Monoclonal Antibody

LY6G6D Recombinant Monoclonal Antibody for ELISA, Flow

G6D, also known as Lymphocyte antigen 6 complex locus G6D (LY6G6D), is a type I transmembrane protein belonging to the immunoglobin (Ig) superfamily, that is comprised of cell-surface proteins involved in the immune system and cellular recognition. It is a disulfide linked homodimer with an Ig-like V-type domain and is known to interact with GRB2 and GRB7 in a phosphorylation-dependent manner thus playing an important role in their downstream signal transduction pathways.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LY6G6D
Applications ELISA, Flow Cytometry, Neutralization
Classification Recombinant Monoclonal
Clone 8I8
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene LY6G6D
Gene Accession No. O95868
Gene Alias C6orf23; DADB-110M10.6; G6D; LY6-D; LY6G6D; Lymphocyte antigen 6 complex locus protein G6d; lymphocyte antigen 6 complex, locus G6D; lymphocyte antigen 6 family member G6D; lymphocyte antigen-6 G6D; megakaryocyte-enhanced gene transcript 1 protein; MEGT1; NG25; Protein Ly6-D
Gene Symbols LOC102119090, LY6G6D
Host Species Human
Immunogen Recombinant Human LY6G6D protein.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 102119090, 58530
Target Species Human, Monkey
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.