Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LY6G6F Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501735110 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5110 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5110 Supplier Proteintech Group Inc Supplier No. 258721AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

May play a role in the downstream signal transduction pathways involving GRB2 and GRB7.

Specifications

Antigen LY6G6F
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 0.43 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene LY6G6F
Gene Accession No. Q5SQ64
Gene Alias C6orf21, G6f, LY6G6D, LY6G6F, NG32
Gene Symbols LY6G6F
Host Species Rabbit
Immunogen LY6G6F Fusion Protein Ag22467
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 259215
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.