Learn More
Invitrogen™ LYK5 Polyclonal Antibody

Description
The amino acid sequence used is 100% homologous in Yellow Fluorescent Protein (YFP) and Cyan Fluorescent Protein (CFP).
Specifications
Specifications
| Antigen | LYK5 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA and 0.05% sodium azide |
| Gene | STRADA |
| Gene Accession No. | Q3UUJ4, Q7RTN6 |
| Gene Alias | 2610019A05Rik; 6030402H20Rik; AI480680; E130112C09Rik; LYK5; NY-BR-96; PMSE; protein kinase LYK5; serologically defined breast cancer antigen NY-BR-96; STE20-like pseudokinase; STE20-related adapter protein; STE20-related kinase adapter protein alpha; STE20-related kinase adaptor alpha; Stlk; Strad; STRAD alpha; Strada |
| Gene Symbols | STRADA |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.