Learn More
MP Biomedicals™ LymphoSpinner™ Lymphocyte Isolation Tubes
Catalog No. 091692425 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Volume (Metric) Working:
15 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Working
09-169-2425 15 mL
09-169-2525 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 09-169-2425 Supplier MP Biomedicals™ Supplier No. 091692425
Add to Cart
Add to Cart
An innovative separation tube with a porous membrane for optimal separation of lymphocytes and mononuclear cells from blood and tissues.
  • Biologically inert barrier prevents mixture of the sample with the Lymphocyte Separation Medium (LSM™)
  • Maintains discontinuous gradient and improves separations with higher yield and less processing time
  • Sterile and designed for single use only
TRUSTED_SUSTAINABILITY
Specifications
Product Type Isolation Tube
Quantity 25 Tubes
Volume (Metric) Working 15 mL
Sterility Sterile