Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PeproTech Lymphotactin Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Catalog No. p-7244950
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-271-8490 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-271-8490 Supplier PeproTech Supplier No. 500P40BT500UG
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AA Sequence of recombinant protein: GSEVSDKRTC VSLTTQRLPV SRIKTYTITE GSLRAVIFIT KRGLKVCADP QATWVRDVVR SMDRKSNTRN NMIQTKPTGT QQSTNTAVTL TG. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human Lymphotactin (XCL1). Anti-Human Lymphotactin (XCL1)-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect hLymphotactin by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human Lymphotactin (500-P40) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant hLymphotactin. Western Blot: To detect hLymphotactin by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hLymphotactin is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P40BT-1MG will be provided as 2 x 500 μg

Lymphotactin, otherwise known as XCL1, is a member of the C-chemokine family that uniquely acts as a chemoattractant for lymphocytes, but not monocytes or neutrophils, suggesting an important regulatory role in lymphocyte trafficking and inflammation. Lymphotactin, shown to signal through chemokine XC receptor 1 (XCR1), is detected at the highest level in the spleen and at low levels in the lungs, small intestine and colon, and is abundantly and selectively expressed by activated T cells, in particular CD8+ T cells, and is also produced by NK cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lymphotactin
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.1-1.0 mg/mL
Conjugate Biotin
Formulation PBS with no preservative
Gene Xcl1
Gene Accession No. P47992
Gene Alias AI661682; ATAC; C motif chemokine 1; chemokine (C motif) ligand 1; cytokine SCM-1; Lptn; LTN; Lymphotactin; lymphotaxin; SCM1; SCM-1; SCM1A; SCM-1a; SCM1alpha; SCM-1alpha; SCM-1-alpha; SCM1beta; SCM-1beta; SCM-1-beta; SCYC1; single cysteine motif 1a; small inducible cytokine subfamily C, member 1 (lymphotactin); small-inducible cytokine C1; XC chemokine ligand 1; X-C motif chemokine ligand 1; XCL1
Gene Symbols Xcl1
Host Species Rabbit
Immunogen E.coli-derived Recombinant Human Lymphotactin (XCL1).
Purification Method Antigen affinity chromatography
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6375
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.