Learn More
Invitrogen™ Lyn Polyclonal Antibody

Description
Recommended positive controls: HeLa, CD44-knockout HeLa, Raji, K562, THP-1. Predicted reactivity: Mouse (94%), Rat (95%), Xenopus laevis (83%), Sheep (87%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | Lyn |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | LYN |
| Gene Accession No. | P07948, P25911, Q07014 |
| Gene Alias | AA407514; CD180; CD180 antigen; CD180 molecule; CH73-38P6.3; FLJ26625; Hck-2; JTK8; lck/Yes-related novel protein tyrosine kinase; LY64; Ly78; lymphocyte antigen 64; lymphocyte antigen-64, radioprotective, 105kDa; LYN; lyn protein non-receptor kinase; lyn protein tyrosine kinase; LYN proto-oncogene, Src family tyrosine kinase; p53Lyn; p56Lyn; Radioprotective 105 kDa protein; RP105; Tyrosine-protein kinase Lyn; v-yes-1 Yamaguchi sarcoma viral related oncogene homolog; v-yes-1 Yamaguchi sarcoma viral related oncogene protein-like protein; Yamaguchi sarcoma viral (v-yes-1) oncogene homolog; zgc:92124 |
| Gene Symbols | LYN |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.