Learn More
Sartorius LYOPHOLIZED MOUSE IGG2A FABFLU
Supplier:  Sartorius 4750
Encompass
Lyopholized Mouse IgG2a FabFluor-pH Antibody Labeling Reagent (50 µg) for labeling antibodies containing Mouse IgG2a Fc region
Catalog No. NC1750348
Add to Cart
Description
Description
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov