Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LYPD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA526749 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA526749 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA526749 Supplier Invitrogen™ Supplier No. PA526749
  • $616.00
    $489.65 / Each of 1

    Save $126.35

    21% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

LYPD1 Polyclonal Antibody for IHC (P), Western Blot

Believed to act as a modulator of nicotinic acetylcholine receptors (nAChRs) activity. In vitro increases receptor desensitization and decreases affinity for ACh of alpha-4:beta-2- containing nAChRs. May play a role in the intracellular trafficking of alpha-4:beta-2 and alpha-7-containing nAChRs and may inhibit their expression at the cell surface. May be involved in the control of anxiety.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LYPD1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.35 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene LYPD1
Gene Accession No. Q8BLC3, Q8N2G4
Gene Alias 2700050C12Rik; AI853408; C530008O16Rik; Ly-6/neurotoxin-like protein 2; LY6/PLAUR domain containing 1; ly6/PLAUR domain-containing protein 1; Lynx2; LYPD1; Lypdc1; MNCb-0671; PHTS; PSEC0181; Putative HeLa tumor suppressor; UNQ3079/PRO9917
Gene Symbols LYPD1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 93-122 amino acids from the C-terminal region of human LYPD1.
Purification Method Protein A, Antigen affinity chromatography
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116372, 72585
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.