Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quidel™ Lyra Direct SARS-CoV-2 Molecular Assay
SDP

Catalog No. 23044028 Shop All QuidelOrtho Products
Change view
Click to view available options
Includes:
Nasal/oropharyngeal swabs, transport tubes, deep well plates
Nasal/oropharyngeal swabs, transport tubes, microfuge tubes
Nasopharyngeal swabs, transport tubes, deep well plates
Nasopharyngeal swabs, transport tubes, microfuge tubes
Tests and Controls
Sample Type:
Nasal, Nasopharyngeal, Oropharyngeal Swab
Nasal/oropharyngeal Swab
Nasopharyngeal Swab
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes Sample Type
23-044-028 Nasal/oropharyngeal swabs, transport tubes, microfuge tubes Nasal/oropharyngeal Swab
23-044-012 Tests and Controls Nasal, Nasopharyngeal, Oropharyngeal Swab
23-044-029 Nasopharyngeal swabs, transport tubes, microfuge tubes Nasopharyngeal Swab
23-044-030 Nasal/oropharyngeal swabs, transport tubes, deep well plates Nasal/oropharyngeal Swab
23-044-031 Nasopharyngeal swabs, transport tubes, deep well plates Nasopharyngeal Swab
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 23-044-028 Supplier Quidel™ Supplier No. M942
Add to Cart
Add to Cart
This item is not returnable. View return policy

For in vitro qualitative detection of human coronavirus SARS-CoV-2 from viral RNA extracted from nasal, nasopharyngeal (NP) or oropharyngeal (OP) swab specimens from patients with signs and symptoms of COVID-19, Assay targets non-structural Polyprotein (pp1ab) of SARS-CoV-2 virus

  • RT-PCR
  • One-step reagent setup
  • Results in <70 minutes after extraction
  • No freezer necessary
  • Refrigration storage

Specifications

Format Microfuge
Sample Type Nasal/oropharyngeal Swab
Type SARS-CoV-2 Assay
For Use With (Equipment) Emergency Use Authorization (EUA) for use on Applied Biosystems™ 7500 Fast Dx, Applied Biosystems 7500 Standard, Roche LightCycler™ 480 Instrument II, Roche cobas™ z 480, Qiagen Rotor-Gene™ Q, Bio-Rad CFX96 Touch™, Thermo Fisher QuantStudio™ 7 Pro
Includes Nasal/oropharyngeal swabs, transport tubes, microfuge tubes
Quantity 96 Tests, 100 Microfuge Tubes (1.5 mL), 100 Swabs, 100 Transport Tubes
Reagent Type Viral transport media
Sterility Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.