Learn More
Thermo Scientific™ Lys-Bradykinin Acetate Salt
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Purification Method | HPLC |
| Common Name | Kallidin |
| Content And Storage | Store at -20°C |
| Format | Powder |
| For Use With (Application) | Endogenous bradykinin receptor agonist |
| Molecular Weight (g/mol) | 1188.40 |
| Product Type | Lys-Bradykinin acetate salt |
| Quantity | 5 mg |
| Regulatory Status | RUO - research use only |
| Sequence | Lys-Arg-Pro-Pro-Gly- Phe-Ser-Pro-Phe-Arg |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.