Learn More
CPC Scientific H-Ala-Arg-Thr-Lys-Gln-Thr-Ala-Arg-Lys-Ser-Thr-Gly-Gly-Lys(Ac)-Ala-Pro-Arg-Lys-Gln-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific HIST025A
Encompass
This peptide is the fragment of Histone H3 (1-19) acetylated at Lys14.ONE-LETTER SEQUENCE: ARTKQTARKSTGG-K(Ac)-APRKQMOLECULAR FORMULA: C87H159N35O26MOLECULAR WEIGHT:2111.42STORAGE CONDITIONS: -20 5CRESEARCH AREA: MiscellaneousREFERENCES: Borra, MT. et al. Biochem. 43, 9877 (2004)
Catalog No. 50-210-9458
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.