Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Lys20/Lys21 Monoclonal Antibody (31F5)
GREENER_CHOICE

Catalog No. 01670294
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-670-294 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-670-294 Supplier Invitrogen™ Supplier No. MA110037
  • $599.00
    $497.30 / Each of 1

    Save $101.70

    17% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-10037 detects Lys20 (MW 47.1kDa) and Lys21 (MW 48.6kDa) from S. cerevisiae samples, which appears as a closely-spaced doublet on Western blots.

Lys20 and Lys21 are two isozymes of yeast homocitrate synthase. Both proteins are localized to the nucleoplasm of the nucleus and share 90% homology.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lys20/Lys21
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 31F5
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Tissue culture supernatant with 5mM sodium azide
Gene LYS21
Gene Accession No. P48570, Q12122
Gene Alias D1298; D2195; HCS; homocitrate synthase LYS20; homocitrate synthase LYS21; Homocitrate synthase, cytosolic isozyme; Homocitrate synthase, mitochondrial; LYS20; LYS21; YDL131W; YDL182W
Gene Symbols LYS20, LYS21
Host Species Mouse
Immunogen Yeast nuclear preparation.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 851346, 851425
Target Species Yeast
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.