Learn More
CPC Scientific H-Ala-Thr-Lys-Ala-Ala-Arg-Lys(Ac)-Ser-Ala-Pro-Ser-Thr-Gly-Gly-Val-Lys-Lys-Pro-His-Arg-Tyr-Arg-Pro-Gly-Gly-Lys(Biotin)-NH2 (trifluoroacetate salt) 0.25MG
SDP
Supplier:  CPC Scientific HIST051A
Encompass
This peptide is the fragment of Histone H3 (21-44) acetylated at Lys 27 with an additional C-terminal Gly followed by a biotinylated Lys. Acetylation of histone H3 at Lys 27 is associated with many active mammalian genes. In Arabidopsis thaliana, the acetylated histone at Lys 27 is nontransposable element gene-specific.ONE-LETTER SEQUENCE: ATKAAR-K(Ac)-SAPSTGGVKKPHRYRPG-GK(Biotin)-NH2MOLECULAR FORMULA: MOLECULAR WEIGHT:2974.5STORAGE CONDITIONS: -20 5CRESEARCH AREA: MiscellaneousREFERENCES: Charron, JB. et al. Plant Cell 21, 12 (2009); Tie, F. et al. Development 136, 18 (2009)
Catalog No. 50-210-9493
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.