Learn More
CPC Scientific Biotin-Lys-Gly-Gly-His-Leu-Lys-Ser-Lys(Me2)-Lys-Gly-Gln-Ser-Thr-Ser-Arg-His-Lys-Lys-Leu-Met-Phe-Lys-Thr-Glu-Gly-Pro-Asp-Ser-Asp-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific CANC012B
Encompass
This is a biotin-labeled and amidated peptide derived from residues 368-393 of human p53 tumor suppressor protein with dimethylation at Lys372 and additional KGW residues on its N-terminus. Methylation at Lys372 activates p53 and induces upregulation of target gene p21. Methylation at Lys372 also inhibits Lys370 methylation by interfering with Smyd2, a lysine methyltransferase, and p53 interaction. Lys 370 methylation reduces p53 transcriptional targets.ONE-LETTER SEQUENCE: Biotin-KGGHLKS-K(Me2)-KGQSTSRHKKLMFKTEGPDSD-NH2MOLECULAR FORMULA: MOLECULAR WEIGHT:3467STORAGE CONDITIONS: -20 5CRESEARCH AREA: CancerREFERENCES: Huang, J. et al. Nature 440, 629 (2006)
Catalog No. 50-210-8887
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.