Learn More
CPC Scientific Biotin-Lys-Gly-Trp-His-Leu-Lys-Ser-Lys(Me1)-Lys-Gly-Gln-Ser-Thr-Ser-Arg-His-Lys-Lys-Leu-Met-Phe-Lys-Thr-Glu-Gly-Pro-Asp-Ser-Asp-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific CANC009B
Encompass
This is human tumor suppressor protein p53 (368-393) monomethylated at Lys373 and biotinylated through a KGW linker at its N-terminus. Methylation at Lys373 is catalyzed by G9a and Glp lysine methyltransferases and is associated with inactive p53. ONE-LETTER SEQUENCE: Biotin-KGWHLKS-K(Me1)-KGQSTSRHKKLMFKTEGPDSD-NH2MOLECULAR FORMULA: MOLECULAR WEIGHT:3582.2STORAGE CONDITIONS: -20 5CRESEARCH AREA: CancerREFERENCES: Krishnan, S. et al. ChemBioChem 12, 254 (2011); Huang, J. et al. J Biol Chem 285, 9636 (2010)
Catalog No. 50-210-8882
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.