Learn More
CPC Scientific Biotin-Lys-Gly-Gly-His-Leu-Lys-Ser-Lys-Lys(Ac)-Gly-Gln-Ser-Thr-Ser-Arg-His-Lys-Lys-Leu-Met-Phe-Lys-Thr-Glu-Gly-Pro-Asp-Ser-Asp-NH2 (trifluoroacetate salt) 0.25MG
SDP
Supplier:  CPC Scientific CANC002A
Encompass
This peptide is tumor suppressor protein, p53 (368-393). There is a biotinylated lyine followed by a GG linker at the N-terminal and an acetylated lysine at lysine 373. Activation of the p53 protein results in the transcription of antiproliferative proteins and induction of apoptosis. The p53 tumor suppressor is a principal mediator of growth arrest, senescence, and apoptosis in response to a broad array of cellular damage.ONE-LETTER SEQUENCE: Biotin-KGGHLKSK-K(Ac)-GQSTSRHKKLMFKTEGPDSD-NH2MOLECULAR FORMULA: MOLECULAR WEIGHT:3481STORAGE CONDITIONS: -20 5CRESEARCH AREA: CancerREFERENCES: Kanovsky, M. et al. Proc Natl Acad Sci 98, 12438 (2001)
Catalog No. 50-210-8868
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.