Learn More
CPC Scientific Biotin-LC-Lys-Lys-Gly-Gln-Ser-Thr-Ser-Arg-His-Lys-Lys(Ac)-Leu-Met-Phe-Lys-Thr-Glu-Gly-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific CANC007B
Encompass
This is an amidated peptide derived from amino acid residues 372-389 of human p53 tumor suppressor protein with acetylation at Lys382. It is biotinylated at its N-terminus through a 6-carbon LC linker. Activated p53 functions to induce cell cycle arrest and apoptosis in response to signals such as DNA damage. Acetylation of p53 reduces ubiquitination by MDM2 and increases p53 half-life in vivoONE-LETTER SEQUENCE: Biotin-LC-KKGQSTSRHK-K(Ac)-LMFKTEG-NH2MOLECULAR FORMULA: MOLECULAR WEIGHT:2472STORAGE CONDITIONS: -20 5CRESEARCH AREA: CancerREFERENCES: Kubbutat, M. et al. Lett. Nature 387, 299 (1997); Li, M. et al. J. Biol. Chem. 277, 50607 (2002)
Catalog No. 50-210-8879
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.