Learn More
CPC Scientific H-Thr-Ser-Arg-His-Lys-Lys(Me3)-Leu-Met-Phe-Lys-Thr-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific CANC015B
Encompass
p53K382Me3 is derived from amino acid residues 377-387 of human p53 tumor suppressor protein, with trimethylation at Lys382. Trimethlyation of K382 does not occur in vivo but can be used as a control. ONE-LETTER SEQUENCE: TSRHK-K(Me3)-LMFKTMOLECULAR FORMULA: MOLECULAR WEIGHT:1418.7STORAGE CONDITIONS: -20 5CRESEARCH AREA: CancerREFERENCES: irskaia, I. et al. J Biol Chem 283, 34660 (2008); Shi, X. et al. Mol Cell 27, 636 (2007)
Catalog No. 50-210-8893
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.