Learn More
CPC Scientific H-Glu-Ile-Ala-Gln-Asp-Phe-Lys(Ac)-Thr-Asp-Leu-Arg-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific HIST018A
Encompass
This peptide is the fragment of histone H3 (73-83) with acetylation at Lys79. Histone acetylation generally is associated with transcriptional activation. Sirt2, a NAD+ dependent lysine deacetylase, preferentially deacetylates Histone H3 at Lysine 79.ONE-LETTER SEQUENCE: EIAQDF-K(Ac)-TDLRMOLECULAR FORMULA: C60H96N16O21MOLECULAR WEIGHT:1377.51STORAGE CONDITIONS: -20 5CRESEARCH AREA: MiscellaneousREFERENCES: Bheda, P. et al. PNAS 109, E916 (2012)
Catalog No. 50-210-9449
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.