Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Lysis/Binding Buffer for Dynabeads™ mRNA Purification Kits

Catalog No. A33562
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 x 60 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A33562 1 x 60 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A33562 Supplier Invitrogen™ Supplier No. A33562
Add to Cart
Edge
Add to Cart

This Lysis/Binding Buffer is included in Dynabeads™ mRNA Purification kits.

This Lysis/Binding Buffer is included in the following Dynabeads™ mRNA Purification kits (Cat. No. 61011, 61012, and 61021). It is made available separately for applications that require more Lysis/Binding Buffer than is provided in the kit.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 1 x 60 mL, store (at 2°C to 8°C)
Product Line Dynabeads
Quantity 1 x 60 mL
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.