Learn More
Apexbio Technology LLC CD38 inhibitor 1(Synonyms: 78c, CD38-IN-78c, CD38 inhibitor 78c, CD38 selective inhibitor), 50mg, CAS: 1700637-55-3.
SDP
Supplier:  Apexbio Technology LLC B875250
Encompass_Preferred
CD38 inhibitor 1 (CAS 1700637-55-3) is a potent selective small molecule inhibitor targeting CD38 enzymatic activity in human and mouse systems Reported IC50 values are approximately 7 3 nM for human CD38 and 1 9 nM for mouse CD38 CD38 is implicated in intracellular calcium signaling and NAD metabolic regulation Blocking CD38 enzymatic function enables exploration of its biological roles and potential therapeutic relevance in immunological processes inflammation autoimmune disorders and cancer metabolism through both in vitro and in vivo studies
Catalog No. 50-000-60729
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.