Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Lysobacter Enzymogenes Arg-C Recombinant Protein, PeproTech®

Catalog No. 450545UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μg
5 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
450-54-5UG 5 μg
450-54-20UG 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 450-54-5UG Supplier Gibco™ Supplier No. 450545UG
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

450-54-100UG will be provided as multiple 5 μg (450-54-5UG) and 20 μg (450-54-20UG) vials. Recombinant Lysobacter Enzymogenes Arg-C is a 26.8 kDa protease consisting of 252 amino acid residues including a C-terminal His-Tag. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Proteases (also called Proteolytic Enzymes, Peptidases, or Proteinases) are enzymes that hydrolyze the amide bonds within proteins or peptides. Most proteases act in a specific manner, hydrolyzing bonds at, or adjacent to specific residues, or a specific sequence of residues contained within the substrate protein or peptide. Proteases play an important role in most diseases and biological processes, including prenatal and postnatal development, reproduction, signal transduction, the immune response, various autoimmune and degenerative diseases, and cancer. They are also an important research tool, frequently used in the analysis and production of proteins. Arg-C specifically cleaves at the carboxyl side of Arginine residues. Arg-C has a sulfhydryl requirement; it is activated by dithiothreitol, cysteine, or other sulfhydryl-containing reagents. The presence of calcium ions is essential. The enzyme is inhibited by oxidizing agents and sulfhydryl reactants, and by Co^2+, Cu2+, Cd^2+, and heavy metal ions.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Functional Assay
Formulation protein with no preservative
Molecular Weight (g/mol) 26.8 kDa
Name Lysobacter Enzymogenes Arg-C
Quantity 5 μg
Source Insect cells
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Common Name Arg-C
Biological Activity The reaction is measured as an increase in absorbance at 253 nm resulting from the hydrolysis of N-benzoyl-L-arginine ethyl ester (BAEE).
Conjugate Unconjugated
Recombinant Recombinant
Protein Tag His-tag
Sequence GVGDIGSSDY CEKDIVCRVK PSAEFLSASK SVARMVFTPK TGYTGYCSGT LLNNSNSPKR QLFWSAAHCI STQKVANTLQ TYWLYDATGC DNDTLSDKAV TLTGGATLLH SHATRDTLLL ELKSAPPSGA YYAGWNSSAI ATKGTAIEGI HHPSGDLKKY SLGSVTALSS TIDGKKPLTK VAWTTGVTEG GSSGSGLFTI SSTSGYQLRG GLYGGTSYCS APSDPDYYSQ LDGVWSSIKT YFSPHHHHHH HH
Content And Storage -20°C
Expression System Insect cells
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.