Learn More
Market Lab Lysol IC Quaternary Disinfectant Cleaner, 1 Gallon
Supplier:  Market Lab 60282
Encompass_Preferred
Lysol IC Quaternary Disinfectant Cleaner is a concentrated, hospital-grade disinfectant that kills 99.9% of germs. It can be diluted at 1:64 ratio for effective cleaning and disinfecting. This EPA-registered disinfectant cleaner is designed for use on hard, nonporous surfaces in healthcare settings. It cleans quickly by removing dirt, grime, blood, urine, fecal matter and other organic matter commonly found in healthcare facilities. This Lysol concentrate requires no rinsing and leaves no residue after use. It meets OSHA bloodborne pathogen standards. The active ingredients are alkyl dimethyl benzyl ammonium chlorides and alkyl dimethyl ethylbenzyl ammonium chlorides.
Catalog No. NC2601431
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.