Learn More
Bioworld LYSOSTAPHIN (RECOMBINANT)
Supplier:  Bioworld 220605361
Encompass_Preferred
Lysostaphin, an endopeptidase specific for the cell wall peptidoglycan of staphylococci, is an extremely potent anti-staphylococcal agent. Physical Appearance: Sterile-filtered lyophilized powder. When resuspended, it is a clear solution. Recommended reconstitution: At a concentration of 10mg/ml in 10-20mM sodium acetate, pH 4.5, to be further diluted as necessary. Recommended reaction buffer is 200mM Tris-HCl pH 8. Pkg yield (10mg) Size (1mg)
Catalog No. 50-253-634
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.