Learn More
Diagnostic Biosystems LYSOZYME Lysozyme
Supplier:  Diagnostic Biosystems RP02805
Encompass_Preferred
This antibody reacts with lysozyme. It stains granulocytes, monocytes and macrophages in human tonsil, skin and colon. This antibody does not crossreact with any other cell types.
Catalog No. 50-238-5488
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.