Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lysozyme, chicken egg white

Catalog No. p-7041914
Change view
Click to view available options
Quantity:
1g
5g
25g
100 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6070103 1g
AAJ60701-22 100 g
AAJ6070106 5g
AAJ6070114 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAJ6070103 Supplier Thermo Scientific Chemicals Supplier No. J6070103
Only null left
Add to Cart

Lysozyme, chicken egg white, CAS # 12650-88-3, is an enzyme that catalyzes the hydrolysis of peptidoglycans found in gram-positive bacterial cell walls, causing bacterial cell lysis. | CAS: 12650-88-3 | C125H196N40O36S2

Catalyzes the hydrolysis of peptidoglycans found in bacterial cell walls. Lysozyme catalyzes is utilized for the hydrolysis of peptidoglycans found in bacterial cell walls. It is used as a precursor in the production of spheroplasts. It is also used as a preservative against many food spoiling microorganisms. It is used in skin care in order to cure and prevent acne and bed sores, dental and oral conditions. It is used for lysing E. coli and Streptomycetes for extraction purposes such as extracting group specific antigen. Further, it is used in non-pasteurized beer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Lysozyme, chicken egg white is a naturally occurring enzyme that hydrolyzes the peptidoglycans in the bacterial cell wall, resulting in cell wall lysis

Application

• Lysozyme is utilized for the hydrolysis of peptidoglycans found in bacterial cell walls
• It is used as a precursor in the production of spheroplasts in laboratory settings
• It is used for lysing E. coli and Streptomycetes for extraction purposes, such as extracting group-specific antigen

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12650-88-3
Molecular Formula C125H196N40O36S2

Specifications

Color White
Solubility Soluble in water at 10mg/ml.
Content And Storage Store at -20°C
Quantity 1g
Product Type Lysozyme
Form Powder

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.