Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Lysozyme (Egg White)

Catalog No. 89951334 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-951-334 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-951-334 Supplier Abnova™ Supplier No. P5237
Add to Cart
Add to Cart

Used for Enzyme

Specifications

Accession Number P00698
For Use With (Application) Enzyme Activity
Gene ID (Entrez) 396218
Molecular Weight (g/mol) 14.3kDa
Name Lysozyme (Egg White)
Preparation Method Native protein purified from Egg Whites
Quantity 1 g
Storage Requirements Store at 4°C.
Regulatory Status RUO
Gene Alias LYZC
Common Name LYZ
Gene Symbol LYZ
Assays Lysozyme Assay; Nucleic acid preparation; Protein purification from inclusion bodies; Plasmid preparation (to break down membranes and cell wall); Hydrolysis of chitin, Lysozyme Assay; Hydrolysis of bacterial cell walls.
Biological Activity ≥5,000 units per mg dry weight. One Unit is equal to a decrease in turbidity of 0.001 per minute at 450nm at pH 7.0 and 25°C, using a 0.3mg/ml suspension of Micrococcus lysodeikticus cells as substrate.
Species Chicken
Protein Tag None
Expression System Native protein purified from Egg Whites
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.