Learn More
Alpco Ltd Lysozyme ELISA
SDP
Supplier:  Alpco Ltd 30LYSHUE01
Encompass
The Lysozyme ELISA is intended for the quantitative determination of lysozyme in human stool samples.Compatible with the Easy Stool Extraction DeviceSpecifically validated for use exclusively with stool samples
Catalog No. 50-922-055
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.