Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Lysozyme Monoclonal Antibody (A5A9)
GREENER_CHOICE

Catalog No. PIMA537804
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537804 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537804 Supplier Invitrogen™ Supplier No. MA537804
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive controls: HL-60 cell lysates, human spleen tissue, human stomach carcinoma tissue, A549.

This gene encodes human lysozyme, whose natural substrate is the bacterial cell wall peptidoglycan (cleaving the beta[1-4]glycosidic linkages between N-acetylmuramic acid and N-acetylglucosamine). Lysozyme is one of the anti-microbial agents found in human milk, and is also present in spleen, lung, kidney, white blood cells, plasma, saliva, and tears. Missense mutations in LYZ have been identified in heritable renal amyloidosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lysozyme
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone A5A9
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene LYZ
Gene Accession No. P61626
Gene Alias 1,4-beta-N-acetylmuramidase C; 1700038F02Rik; 4-beta-N-acetylmuramidase C; AI326280; Allergen Gal d IV; bA534G20.1; c-type lysozyme; dystrophin; egg white lysozyme; Gal d 4; KAAG648; LYC2; Lys; Lysm; lysozyme; lysozyme (renal amyloidosis); lysozyme 1; lysozyme 2; lysozyme C; Lysozyme C type M; lysozyme C type P; Lysozyme C, spleen isozyme; lysozyme C-1; Lysozyme C-2; lysozyme C-3; lysozyme d1; lysozyme F1; lysozyme like 1; lysozyme-like 1; lysozyme-like protein 1; lysozyme-like protein 2; Lysz; LYZ; Lyz1; Lyz2; LYZC; LYZD1; LYZF1; LYZL1; Lyzs; Lzm; Lzm-s1; Lzp; Lzp-s; P lysozyme structural; PRO1278; renal amyloidosis; RGD1559869; unnamed protein product; UNQ648/PRO1278
Gene Symbols LYZ
Host Species Mouse
Immunogen Recombinant protein within Human Lysozyme aa 1-141.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4069
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.