Learn More
Description
Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human, Baboon, Monkey, Pig, Cat and Dog. Does not react with mouse and rat. Others not known.
Epitope: Not determined
Immunogen: Human lysozyme purified from the urine of patients with monocytic leukemia
Positive Control: Tonsil, colon or skin
Cellular Localization: Cytoplasmic
Ab-1 was adsorbed with human plasma and urine proteins to remove the cross-reacting antibodies. Ab-1 labels myeloid cells, histiocytes, granulocytes, macrophages, and monocytes. This antibody is helpful in the identification of myeloid or monocytic nature of acute leukemia.
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Specifications
| Antigen | Lysozyme/Muramidase Ab-1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Clone | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Immunogen | Human lysozyme purified from the urine of patients with monocytic leukemia |
| Quantity | 500 μL |
| Regulatory Status | IVD |
| Research Discipline | Enzyme |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.