Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lysozyme Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501729909 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-9909 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-9909 Supplier Proteintech Group Inc Supplier No. 150131AP150UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Lysozyme is synthesized predomitly in reactive histiocytes rather than in resting, unstimulated phagocytes.

Specifications

Antigen Lysozyme
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.15 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene LYZ
Gene Accession No. P17897, P61626, Q05820
Gene Alias 1,4 beta N acetylmuramidase C, lysozyme, lysozyme (renal amyloidosis), Lysozyme C, LYZ, LZM
Gene Symbols LYZ, Lyz1, Lyz2
Host Species Rabbit
Immunogen Lysozyme Fusion Protein Ag6917
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17110, 25211, 4069
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.