Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

m-Cresol, ≥99% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019527 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9527 5 mL
11-102-0671 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-9527 Supplier MilliporeSigma Supelco Supplier No. 659965MLF
Add to Cart
Add to Cart

m-Cresol is an isomer form of Cresol and is mainly used as an intermediate in the manufacture of chemicals. It is toxic in nature. It is a component of coal tar creosote which is used for wood preservation. It also possess bactericide and fungicide property.

Featured Industries: Agriculture, Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 108-39-4
Ignition Point 1036°F
Vapor Pressure <1 mmHg (20°C)
Vapor Density 3.72 (vs. air)
Density 1.034 g/mL (at 25°C)
Flash Point 86°C
Physical Form Neat
Linear Formula CH3C6H4 OH
MDL Number MFCD00002302
Melting Point 8°C to 10°C
Boiling Point 203°C
Synonym 1-Hydroxy-3-methylbenzene; 3-Hydroxytoluene; 3-Methylphenol
Type Analytical Standard
Quantity 5 mL
Formula Weight 108.14
Percent Purity ≥99% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.