Learn More
Zymo Research Corporation EZ-96 DNA Methylation™ Kit (Shallow-Well) (2 x 96 Rxns)
SDP
Supplier:  Zymo Research Corporation D5003
Encompass_Preferred
The EZ-96 DNA MethylationTM Kit is based on the three-step reaction that takes place between cytosine and sodium bisulfite where cytosine is converted into uracil. The products innovative desulphonation technology eliminates otherwise cumbersome precipitations.The kit is designed to reduce template degradation, minimize DNA loss during treatment and clean-up, while ensuring complete conversion of the DNA. Purified, converted DNA is ideal for PCR amplification for downstream analyses including endonuclease digestion, sequencing, microarrays, etc.
Catalog No. 50-125-1516