Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

m,m'-Ditolylamine 98.0+%, TCI America™
SDP

Catalog No. D151225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D151225G 25 g
D15125G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D151225G Supplier TCI America Supplier No. D151225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 626-13-1
Molecular Formula C14H15N
Molecular Weight (g/mol) 197.281
MDL Number MFCD00059315
InChI Key CWVPIIWMONJVGG-UHFFFAOYSA-N
Synonym di-m-tolylamine, 3,3'-dimethyldiphenylamine, m,m'-ditolylamine, dim-tolylamine, bis 3-methylphenyl amine, 3-methyl-n-3-methylphenyl aniline, benzenamine, 3-methyl-n-3-methylphenyl, di-m-tolyl-amine, 3,3'-ditolylamine, pubchem8963
PubChem CID 7016139
IUPAC Name 3-methyl-N-(3-methylphenyl)aniline
SMILES CC1=CC(=CC=C1)NC2=CC=CC(=C2)C

Specifications

Melting Point -12°C
Boiling Point 200°C
Quantity 25 g
Formula Weight 197.28
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material m,m′-Ditolylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.