Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

m-Methoxycinnamic Acid, Predominantly trans 97%, Thermo Scientific™

Catalog No. AC125830050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC125830050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC125830050 Supplier Thermo Scientific Chemicals Supplier No. 125830050

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6099-04-3
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.19
InChI Key LZPNXAULYJPXEH-AATRIKPKSA-N
Synonym 3-methoxycinnamic acid, 3-3-methoxyphenyl acrylic acid, m-methoxycinnamic acid, 2e-3-3-methoxyphenyl acrylic acid, 2e-3-3-methoxyphenyl prop-2-enoic acid, trans-3-methoxycinnamic acid, e-3-3-methoxyphenyl acrylic acid, trans-3-3-methoxyphenyl acrylic acid, 2-propenoic acid, 3-3-methoxyphenyl-, 2e, 3-3-methoxyphenyl prop-2-enoic acid
PubChem CID 637668

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 116°C to 119°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 97%
Linear Formula CH3OC6H4CH=CHCO2H
Quantity 5 g
Beilstein 10, 295
Formula Weight 178.19
Percent Purity ≥96.0%
Physical Form Fine Crystalline Powder
Chemical Name or Material m-Methoxycinnamic acid, 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.