Learn More
Genscript Corporation M-MuLV Reverse Transcriptase
Supplier: Genscript Corporation E0005040000
GenScript M-MuLV Reverse Transcriptase (M-MLV) is derived from a cloned region of the pol gene of MMLV and isolated from an E coli strain overexpressing this construct To increase cDNA yields and get a higher percentage of longer transcripts the M-MLV Reverse Transcriptase has been modified with reduced RNase H activity and expressed free of exogenous RNases and other nucleases The enzyme can synthesize a complementary cDNA strand initiating from a primer using RNA as template (cDNA synthesis) making it ideal for a wide range of applications
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.