Learn More
Specifications
Specifications
| Product Type | HRP Immunodetection Kit |
| Quantity | 1 Kit |
| For Use With (Application) | Western Blot, Immunohistochemistry (IHC), Immunocytochemistry/Immunofluorescence |
| Product Line | Novus |
| Species | Mouse |
| Target Specificity | M.O.M. (Mouse-On-Mouse) Elite IgG |
| Content And Storage | Store at 4°C. Do not freeze. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
