Learn More
Medchemexpress LLC m-PEG6-NHS ester | 1449390-12-8 | >97.0% | 421.44 | C18H31NO10 | 250 MG

Supplier: Medchemexpress LLC HY133066250MG
m-PEG6-NHS ester is a non-cleavable 6-unit polyethylene glycol (PEG) N-hydroxysuccinimide (NHS) ester used as a linker in antibody-drug conjugate (ADC) and proteolysis targeting chimera (PROTAC) synthesis, enabling efficient coupling to amine-containing ligands.
- Non-cleavable 6-unit PEG linker suitable for bioconjugation.
- High purity >97.0% to support research applications.
- Available in multiple pack sizes, including 250 MG.
- Molecular weight 421.44 g/mol and chemical formula C18H31NO10.
- Liquid, colorless to light yellow; store at -20°C (pure) or follow solvent storage recommendations.
- Includes datasheet and safety data sheet for handling and storage information.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.